Order Entry
Germany
ContactUsLinkComponent
Lymphocyte Separation Medium
Lymphocyte Separation Medium
Catalog # TNB-4700-L100
Supplier:  Tonbo Biosciences
CAS Number:  
undefined
Lymphocyte Separation Medium
Catalog # TNB-4700-L100
Supplier:  Tonbo Biosciences
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Storage temperature:
    Store undiluted at 2...30 °C
  • Tested applications:
    Flow Cytometry

Specifications

About this item

Lymphocyte Separation Medium (LSM) is a sterile-filtered iso-osmotic solution designed for the in vitro isolation of lymphocytes from diluted whole blood.

The use of LSM is based on the method of isolating lymphocytes using density gradient centrifugation (differential centrifugation), resulting in the formation of several distinct cell layers. Mononuclear cells (lymphocytes and monocytes) and platelets will be contained in the banded plasma-LSM interphase due to their density. Erythrocytes and granulocytes will migrate through the gradient to the bottom of the tube. Lymphocytes can be recovered by aspirating the plasma layer and removing the cells.