Order Entry
Germany
ContactUsLinkComponent
PicoKine Human Caspase 8; ELISA Kit
PicoKine Human Caspase 8; ELISA Kit
Catalog # BSBTEK1426
Supplier:  Boster Bio
CAS Number:  
undefined
PicoKine Human Caspase 8; ELISA Kit
Catalog # BSBTEK1426
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    Caspase-8
  • Description:
    Human Caspase 8 PicoKine; ELISA kit
  • Tests per kit:
    96 wells

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of human Caspase 8.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect Human Caspase 8/CASP8 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatesand serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Caspase 8/CASP8 tissue specificity: Isoform 1, isoform 5 and isoform 7 are expressed in a wide variety of tissues. Highest expression in peripheral blood leukocytes, spleen, thymus and liver. Barely detectable in brain, testis and skeletal muscle.

96wells/kit, with removable strips.