Order Entry
Germany
ContactUsLinkComponent
 
4-Hydroxynonenal (4-HNE) ELISA Kit
  ABCAAB287803-96
 :  Abcam
 :  
undefined
4-Hydroxynonenal (4-HNE) ELISA Kit
  ABCAAB287803-96
 :  Abcam
 :  

 

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Target protein:
    4-hydroxynonenal (4-HNE)
  • Description:
    4-Hydroxynonenal ELISA kit
  • Tests per kit:
    100 assays

 

 

A competitive ELISA kit for the quantitative measurement of 4-hydroxynonenal in biological fluids.

  • Easy, convenient and time-saving method
  • Detection Range: 31,25 to 2000 pg/ml
  • Sensitivity: <18,75 pg/ml
  • Assay precision: Intra-assay: CV <8%, inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed

4-hydroxynonenal (4-HNE) is a lipid peroxidation product derived from oxidised ω-6 polyunsaturated fatty acids such as arachidonic acid. 4-HNE is widely used as a marker of lipid peroxidation.

It is also in higher quantities during oxidative stress due to the increase in the lipid peroxidation chain reaction. It exhibits various biological activities such as cytotoxicity, growth inhibiting activity, genotoxicity, and chemotactic activity.

It also inhibits Sirtuin 3 (SIRT3) via thiol-specific modification.The 4-Hydroxynonenal ELISA kit is a competitive ELISA assay for the quantitative measurement of 4-HNE in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of 4-HNE captured from the samples.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.