Ethyl oleate NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:111-62-6
:85049-36-1
Formula:C₂₀H₃₈O₂
MDL Number:MFCD00009579
:310.52 g/mol
Boiling Pt:190…210 °C
Flash Pt:160 °C
Storage Temperature:Ambient
Specification Test Results
- Assay (As Ethyl (Z)-9-Octadecenoate):To Pass Test
- Presence of Ester:To Pass Test
- Iodine Value:75 - 85
- Peroxide Value:max. 10.0
- Water Determination:max. 1.0 %