Specifications
- Pk:1
- Assay Type:Competitive
- Description:T4 Total ELISA, 96 wells
- Application:Thyroid Function
- Regulatory status:FDA Regulated; CE Approved
Specifications
About this item
An enzyme immunoassay for the quantitative measurement of total thyroxin (T4) in serum or plasma.
- High quality assays with reproducible and reliable results
- Ready-to-use reagents with internal controls
- Very good precision and sensitivity
- Short assay time and incubation steps at room temperature
- Simple and technician friendly tests
Measurement of serum thyroxine concentration is generally regarded as an important in-vitro diagnostic test for assessing thyroid function
The DRG Total T4 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards an antigenic site of the T4 molecule. Endogenous T4 of a patient sample competes with a T4-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is inversely proportional to the concentration of T4 in the sample. After addition of the substrate solution, the intensity of colour developed is inversely proportional to the concentration of T4 in the patient sample.
This importance has provided the impetus for the significant improvement in assay methodology that has occurred in the last three decades. This procedural evolution can be traced from the empirical protein bound iodine (PBI) test to the theoretically sophisticated radioimmunoassay. Not intended for new-born screening.