Specifications
- Pk:20 mL
- Description:Hybridoma cell freezing media
Specifications
About this item
Long-Term Storage of Hybridomas at −80 °C or in liquid nitrogen.
- Up to 3× better recovery rates
- No step-wise freezing
There are certain cell types, such as hybridomas, which exhibit increased sensitivity to ''over'' manipulations. These cells often exhibit higher death rates or unwanted differentiation after long-term term storage and freeze thaw cycling.
It is for these types of cells that Bambanker DIRECT was created to provide tight environmental control. By eliminating the centrifugation step, cells can now be frozen for long-term storage immediately upon addition of the novel cryoprotectant.
Just add Bambanker DIRECT, one-to-one, with your cells in media and directly place in freezer. And like with the original Bambanker, the DIRECT version has no serum, so it is great for cells which animal-derived serum may be an issue.