FAM benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
:64742-49-0
:
:MFCD00282001
:65 °C (1013 hPa)
:0.76 g/cm3 (15 °C)
:–26 °C
:Ambient
:1268
:3,II
:GHS02
:GHS07
:GHS08
:GHS09
- Density (15°C):0.690 - 0.710 kg/m3
- Non-volatile matter (mg/100 ml):max. 2
- Aromatics (as C6H6):max. 0.1%
- Boiling range (60-90 °C) (Vol %):85 - 95%
- n-Hexane:max. 5.0%