Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-chloro-2,4,5-trifluorobenzoate
Methyl 3-chloro-2,4,5-trifluorobenzoate ≥95%
Methyl 3-chloro-2,4,5-trifluorobenzoate ≥95%
Methyl 3-chloro-2,4,5-trifluorobenzoate ≥95%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 3-chloro-2,4,5-trifluorobenzoate ≥95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1214375-02-6
:
Formula:
C₈H₄ClF₃O₂
MDL Number:
MFCD12911498
:
224.56436 g/mol