Order Entry
Germany
ContactUsLinkComponent
Hydrogen peroxide 50%, INTEROX® SG-50
Hydrogen peroxide 50%, INTEROX® SG-50
Catalog # 206983.
Supplier:  SOLVAY
CAS Number:  
Hydrogen peroxide 50%, INTEROX® SG-50
Catalog # 206983.
Supplier:  SOLVAY
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Assay (GC):
    49,0-49,9%
  • stability(1h,96°):
    ≤ 3g/kg*h
  • non-volatile matter (105°):
    <2ppm
  • calcium:
    ≤0,05 ppm
  • iron:
    ≤0,01 ppm
  • aluminium:
    ≤0,05 ppm
  • sodium:
    ≤0,5 ppm
  • chloride:
    ≤0,1 ppm
  • phosphate (PO43-):
    ≤1 ppm
  • sulfate:
    ≤0,1 ppm
  • cadmium:
    ≤0,002 ppm
  • chromium:
    ≤0,002 ppm
  • cobalt:
    ≤0,002 ppm
  • copper:
    ≤0,002 ppm
  • manganese:
    ≤0,002 ppm
  • magnesium:
    ≤0,01 ppm
  • nickel:
    ≤0,002 ppm
  • tin:
    ≤0,002 ppm
  • zinc:
    ≤0,002 ppm
  • silicon:
    ≤0,01 ppm
  • nitrate:
    ≤3 ppm

Specification Test Results

About this item

 :
Boiling Pt:113,9 °C
Density:1,19
UN:2014
ADR:5.1 (8),II
 :34.01 g/mol
EINECS:231-765-0
 :GHS03
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333