Watch Glass Dishes
: Kavalier
Watch glass dishes with fused edges made from soda-lime glass.
- Good chemical and physical properties
- Quick and easy to clean
Not suitable for applications related to strong heat or rapid temperature changes.
: DIN 12341.