Order Entry
Germany
ContactUsLinkComponent
Dispenser for Aura™ Masks
Dispenser for Aura™ Masks
Catalog # 111-2605
Supplier:  3M
undefined
Dispenser for Aura™ Masks
Catalog # 111-2605
Supplier:  3M

Specifications

  • Description:
    Dispenser for 3M Aura™ masks
  • W×D×H:
    260×105×460 mm
  • Pk:
    1

Specifications

About this item

Practical mask dispenser for 3M Aura™ series masks made from durable plastic.

  • Holds 50 Aura™ masks without valve or 25 with valve
  • Quick and simple to load thanks to hinged lid
  • Appropriate for wall mounting
  • Window shows fill level

W×D×H: 26×10,5×46 cm