Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microbiology Test Systems
Antibiotic Tests
Antibiotic assay discs
Antibiotic assay discs
Antibiotic assay discs
Catalog # PANBP06-07100
Supplier:
PAN BIOTECH
CAS Number:
Certificates
Expand image
Antibiotic assay discs
Catalog # PANBP06-07100
Supplier:
PAN BIOTECH
CAS Number:
Certificates
Specifications
Pk:
100 mL
Description:
Penicillin/Streptomycin
Specifications
About this item
Product Details
Documentation
Product Details
Documentation