Specifications
- Weight:1,4 kg
- Temperature range:50...310 °C
- Description:RC 5 single-position hotplate with aluminium top plate
- W×D×H:160×270×85 mm
- Model:RC 5
- Heat output:600 W
- Max. stirring capacity H₂O:Up to 15 L (H₂O)
- Plate dimensions:Ø 135 mm
- Plate material:Aluminium
- Pk:1
Specifications
About this item
Single-position hotplate with resistant aluminum top plate.
- Excellent heating performance up to 310 °C with homogeneous temperature
- The internal electronic parts are protected by the insulating disk which resists even the most challenging environments and applications (IP 42 protection)
- Safety functions to protect users in daily operations, also with analogue models
The RC 5 is equipped with a 135 mm diameter aluminum alloy top plate ensuring a great temperature homogeneity and optimum heat transfer across the entire surface.
It is reliable for everyday laboratory applications such as boiling liquids, basic solution and media preparation as well as dissolving buffers and reagents. The RC 5 enables every laboratory to meet the highest safety needs: Various safety functions, such as safety circuits, over-temperature protection and probe detection alarm, protect the user in daily operations. It is also possible to limit the maximum operating temperature.
Thanks to the insulation of the heating plate and a robust die-cast aluminium housing, all the internal electronic parts are protected, enabling the unit to work even in the most demanding laboratory environments.
The intuitive control panel has been carefully designed with an incline to optimize the user experience and to enable the values shown on the display to be easily read.
Housing material: Aluminum body
Hot plate alarm: Over 50 °C (overtemperature alarm over 330 °C)
Heating control: Analogue