Microplate washer, HydroSpeed™
Catalog # TCAT91200001
Supplier: Tecan
Specifications
- Plate types:96-well plates, 384-well plates
- Wash heads:96 indexing wash head
- Description:Microplate washer for 96- and 384-well plates
- Seq:01
- Model:HydroSpeed
- Dispensing precision:3 - 4 % uniformity across the plate
- Dispensing volume:50 - 400 µl in 50 µl increments (96-well plates)
10 - 120 µl in 10 µl increments (384-well plates) - Priming and rinsing volume:100 - 800 ml
- Residual aspiration volume:<2 µl
- Shaking:Linear in three different intensities
- Wash/waste bottle capacity:2× 4 L wash bottles, 1×10 L waste bottle and 1× 4 L foam trap bottle
- Washing volume:50 - 3000 µl in 50 µl increments (96-well plates)
10 - 1000 µl in 10 µl increments (384-well plates) - Weight:15 kg
- W×D×H:388×430×288 mm
- Pk:1
Specifications
About this item
The HydroSpeed is Tecan’s advanced plate washer, designed for quick, parallel washing of entire 96- and 384-well plates. This avoids time differences between the wells for uniform results. In addition, the HydroSpeed provides full control of wash power for optimized washing of cells, beads and ELISAs.
- Cell protection - extra gentle cell washing with tuneable wash power
- Adjustable wash head positions, speed settings and vacuum level
- Anti-clogging - active prevention of needle clogging
- Easy X-change wash heads for intense cleaning in ultrasonic bath
- Automated buffer switching of four liquids and plate shaking capability
- Predefined plate library and protocols for an easy start
- Plate assistant function for easy teaching of new plates
The HydroSpeed plate washer is ready for ELISA washing, as well as gentle washing of adherent and weakly-adherent cells. It supports soak steps with microplate shaking, as well as buffer switching.
With full on-board operation via the easy-to-use touchscreen interface, the HydroSpeed really does offer you power and control for all your cell, bead and ELISA washing applications.
Ordering information: Accessories not included (must be ordered seperate). Additional configuration available on request.
Delivery: Includes 1× Washer, 1× 10 L set bottle waste, 2× 4 L wash bottles, 1× vacuum pump, 1× power cable, 1× USB stick with manuals in 11 languages.