L(+)-Potassium sodium tartrate tetrahydrate
Catalog # HONE25508.9025
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:282.22 g/mol
Merck Index:13,07755
Melting Pt:70…80 °C
Density:∼1.767 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:206-156-8
CAS Number:6381-59-5
Formula:KOOC(CHOH)₂COONa·4H₂O
MDL Number:MFCD00150989