Order Entry
Germany
ContactUsLinkComponent
(±)-Bromosuccinic acid 96%
(±)-Bromosuccinic acid 96%
Catalog # ABCRAB131969.0100
Supplier:  abcr
CAS Number:  
(±)-Bromosuccinic acid 96%
Catalog # ABCRAB131969.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
Merck Index:14,1437
Boiling Pt:319 °C (1013 hPa)
Melting Pt:164…166 °C (dec.)
Density:2.073 g/cm³ (20 °C)
 :196.99 g/mol
Storage Temperature:Ambient
EINECS:213-087-7
 :GHS07
CAS Number:923-06-8
Formula:HOOCBrCHCH₂COOH
MDL Number:MFCD00004213