Hydrogen peroxide 35% stabilised
Catalog # HONE18304.9060
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:60 kg
- Seq:4463
Specifications
About this item
:
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2014
ADR:5.1,II
:M-Clarity™ quality level = MQ200
:34.01 g/mol
EINECS:231-765-0
:GHS05
:GHS07
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333