Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Hydrofluoric acid 40%

60 kg
Catalog # HONE30112.9060
Toxic
Corrosive
Supplier:   Honeywell Chemicals
Hydrofluoric acid 40%
Catalog # HONE30112.9060
CAS Number:  7664-39-3
Hydrofluoric acid 40%

60 kg

CAS Number:  7664-39-3
Price
Inquire For Price
Log in for availability
1 * 60 kg
Product Details & Documents
  • CAS Number:7664-39-3
  • Formula:HF
  • MDL Number:MFCD00011346
  • Molecular Weight:20.01 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-634-8
  • UN:1790
  • ADR:8,II
Specifications

Specifications

  • Pk:
    60 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.