About this item
The Mag-Bind® cfDNA Kit CE IVD is designed for rapid and reliable isolation of circulating cell-free DNA from 1 to 4 ml plasma/serum samples.
The Mag-Bind® cfDNA Kit CE IVD is designed for rapid and reliable isolation of circulating cell-free DNA from 1 to 4 ml plasma/serum samples. The Mag-Bind® cfDNA Kit CE IVD can be processed manually with 15 ml centrifuge tubes or on automated platforms with appropriate plasticware. The procedure eliminates the need for funnels and vacuum steps providing hands-free operation in automated protocols. The uniquely formulated binding buffer from Omega Bio-Tek allows for large sample volumes to be processed in automated formats with 4 ml serum or plasma being processed in a 24-well plate. The magnetic properties of the Mag-Bind® Particles CH enable fast magnetic separation, especially during steps involving large volumes. The high-binding capability decreases the amount of magnetic particles required thereby reducing the elution volume i.e., cfDNA from up to 4 ml serum or plasma can be eluted in just 50 μl.
Specifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:Biofluids (serum, plasma, saliva)
- Target molecule:DNA