Order Entry
Germany
ContactUsLinkComponent
VWR®, Alternate Metals II, 4 components: Ca 500 mg/l, Na 500 mg/l, K 100 mg/l, Mg 100 mg/l in 2% HNO₃
VWR®, Alternate Metals II, 4 components: Ca 500 mg/l, Na 500 mg/l, K 100 mg/l, Mg 100 mg/l in 2% HNO₃
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Alternate Metals II, 4 components: Ca 500 mg/l, Na 500 mg/l, K 100 mg/l, Mg 100 mg/l in 2% HNO₃
Supplier:  VWR Chemicals
Certificates

Specifications

  • Hazard class description:
    Corrosive
  • UN No.:
    3264
  • Storage temperature:
    Ambient

Specifications

Related Information