Order Entry
Germany
ContactUsLinkComponent
 
VWR®, Bisphenol A bis(diphenyl phosphate) (BADP, BAPP, BPADP, BDP), 100 µg/ml in toluene, CRM
  92340.010
 :  
undefined
VWR®, Bisphenol A bis(diphenyl phosphate) (BADP, BAPP, BPADP, BDP), 100 µg/ml in toluene, CRM
  92340.010
 :  

 

  • Pack type:
    Glass ampoule/vial
  • Pk:
    1 mL
  • Standards:
    CRM ISO 17034
  • CAS No.:
    5945-33-5
  • Description:
    Bisphenol A bis(diphenyl Phosphate) (BADP, BAPP, BPADP, BDP), 100 µg/ml in toluene
  • Storage conditions:
    Ambient (>5 °C)
  • UN No.:
    1294
  • Description:
    Bisphenol A bis(diphenyl phosphate) (BADP,BAPP,BPADP,BDP)

 

 

 : Certified reference material.
 : Highly Flammable.