Ergonomic washable silicone mouse, GETT GCQ®
Artikelnr. GETTKH24207
Supplier: GETT GERATETECHNIK
Specifications
- Description:Silicone computer mouse, white, 105×60×37 mm
- Pk:1
Specifications
About this item
This silicone mouse is a true all-rounder. It can be operated ergonomically and, thanks to the silicone surface, is perfect for application areas with high hygiene requirements.
- Medical grade IP 68 mouse is perfect for applications with high hygiene requirements
- Fully washable and disinfectable
- Silicone cap to protect the USB connector from splashing water during cleaning
- Ergonomic functionality with two mouse buttons as well as 3 scroll buttons
- Laser sensor
- Resolution: 1000 dpi
- Button lifetime: >=2 mio. actuations
- Engineered in Germany
- 2-year warranty on the technical function
This silicone mouse has an ergonomic design and convinces with a pleasant tactile feeling at the highest degree of protection (IP 68). It can therefore be cleaned and disinfected all around. The mouse is a product of the GCQ series from GETT.
Certifications: CE, FCC, REACH, RoHs, IP 68.
Packaging: The mouse is delivered safely packed in brown cardboard box.