Order Entry
Germany
Orders LinkContactUsLinkComponent
Wall mounted scrub sinks

Wall mounted scrub sinks

Supplier:   TEKNOMEK
Wall mounted scrub sinks
Supplier:  TEKNOMEK
Certificates
Wall mounted scrub sinks

8 Options Available Below

Product Details & Documents

Our wall mounted scrub sink is a perfect solution for veterinary practices with a variety of different operations- lever, knee paddle or infra-red sensor

  • Brushed dull polish finish
  • Various spout operations available
  • Battery or mains powered infrared taps with a spout length of 250 mm
  • Lever tap option comes with 150 mm levers
  • Deep bowl to enable scrubbing to elbow
  • Hygienic design to prevent water pooling

Our scrub sink is manufactured from 304 food-grade stainless steel and is available in 1 and 2 stations with a wide range of tap operations; knee operation, battery or mains powered infrared sensors or lever taps.
It comes with a deep bowl and high-up tap enables washing up to the elbow with a tall backsplash provides room for soap dispenser. Perfect for hygiene conscious industries. We supply various add-ons for this sink such as plastic or stainless steel soap dispensers.

Product Family Options

Product Information

  • Availability
    Price
  • TEKMWSSS-1SIB
    Wall mounted scrub sink, one station infra-red battery, fully welded 304-grade stainless steel, 406×500×765 mm
    Wall mounted scrub sink, one station infra-red battery, fully welded 304-grade stainless steel, 406×500×765 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • TEKMWSSS-1SIM
    Wall mounted scrub sink, one station infra-red mains, fully welded 304-grade stainless steel, 406×500×765 mm
    Wall mounted scrub sink, one station infra-red mains, fully welded 304-grade stainless steel, 406×500×765 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • TEKMWSSS-1SKO
    Wall mounted scrub sink, one station knee operated, fully welded 304-grade stainless steel, 406×500×765 mm
    Wall mounted scrub sink, one station knee operated, fully welded 304-grade stainless steel, 406×500×765 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • TEKMWSSS-1SLT
    Wall mounted scrub sink, one station lever taps, fully welded 304-grade stainless steel, 406×500×765 mm
    Wall mounted scrub sink, one station lever taps, fully welded 304-grade stainless steel, 406×500×765 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • TEKMWSSS-2SIB
    Wall mounted scrub sink, two station station infra-red battery, fully welded 304-grade stainless steel, 810×500×765 mm
    Wall mounted scrub sink, two station station infra-red battery, fully welded 304-grade stainless steel, 810×500×765 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • TEKMWSSS-2SIM
    Wall mounted scrub sink, two station infra-red mains, fully welded 304-grade stainless steel, 810×500×765 mm
    Wall mounted scrub sink, two station infra-red mains, fully welded 304-grade stainless steel, 810×500×765 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • TEKMWSSS-2SKO
    Wall mounted scrub sink, two station knee operated, fully welded 304-grade stainless steel, 810×500×765 mm
    Wall mounted scrub sink, two station knee operated, fully welded 304-grade stainless steel, 810×500×765 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • TEKMWSSS-2SLT
    Wall mounted scrub sink, two station lever taps, fully welded 304-grade stainless steel, 810×500×765 mm
    Wall mounted scrub sink, two station lever taps, fully welded 304-grade stainless steel, 810×500×765 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.