About this item
Fully-coated with PVC, EDGE® 14-662 safety gloves ensure greater flexibility and comfort, while protecting hands against chemical, mechanical and biological hazards, in addition to delivering heat and abrasion resistance, and are thereby ideal for workers exposed to oil, grease, acid, caustics and alcohol.
- The EDGE range offers a greater flexibility, when compared to other models in its category
- The largest cost-effectiveness ratio to date in a PVC glove
- Combines excellent protection against several chemical products with mechanical resistance
- Non-slip finish on the palm, fingers, and back of hand for improved grip on wet objects
- Also protects against biological agents
Workers exposed to oil, grease, acid, caustics, and alcohol require reliable protection to keep their hands safe. The rough finish protects against abrasions while offering excellent grip, even in wet and oily conditions. The flexibility and comfort of EDGE gloves lead to less hand fatigue and better worker efficiency.
Recommended for: Chemical processing and preparation; handling dry, wet or oily large objects; maintenance of plant and machines.
Specifications
- Catalog No:
- ANSE14-662/9
- ANSE14-662/10
- ANSE14-662/11
- ANSE14-662/8
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Material:
- PVC
- PVC
- PVC
- PVC
- Finish:
- Fully coated, rough finish
- Fully coated, rough finish
- Fully coated, rough finish
- Fully coated, rough finish
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Palm thickness:
- 1,65 mm
- 1,65 mm
- 1,65 mm
- 1,65 mm
- EN norm:
- EN ISO 374-1:2016 Type C, EN ISO 374-5:2016, EN 388:2016 (3121A)
- EN ISO 374-1:2016 Type C, EN ISO 374-5:2016, EN 388:2016 (3121A)
- EN ISO 374-1:2016 Type C, EN ISO 374-5:2016, EN 388:2016 (3121A)
- EN ISO 374-1:2016 Type C, EN ISO 374-5:2016, EN 388:2016 (3121A)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile