About this item
Durable, touchscreen-compatible gloves that guard against electrostatic discharge.
- ESD certified according to EN 16350 : 2014
- Superior tactile sensitivity
- ZONZ™ Comfort Fit knitwork reduces strain in stress-prone areas
- Nitrile foam coating offers excellent grip and long lasting protection, reducing the total cost of ownership
- Dermatologically approved by Dermatest and certified by OEKO-TEX
The 11-819 series is a safety glove with ESD functionality certified to EN 16350:2014, protecting products against electrostatic discharge. In addition, as part of a grounding chain they can be used in explosion hazard zones. Ultra-thin coating provides increased sensitivity for precision tasks. These gloves also provide long lasting protection in abrasive environments, and are designed to be 20% more breathable than standard safety gloves for cooler, drier hands. Comfort Fit Technology provides a further enhanced fit and feel. Seamless 18 gauge nylon and spandex liner combined with the ultra-thin FORTIX coating provides protection while maintaining bare hand sensitivity for tasks that require dexterity and precision.
Specifications
- Catalog No:
- ANSE11-819/9
- ANSE11-819/8
- ANSE11-819/11
- ANSE11-819/7
- ANSE11-819/10
- ANSE11-819/6
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Finish:
- Palm dipped (Fortix™ Foam Nitrile)
- Palm dipped (Fortix™ Foam Nitrile)
- Palm dipped (Fortix™ Foam Nitrile)
- Palm dipped (Fortix™ Foam Nitrile)
- Palm dipped (Fortix™ Foam Nitrile)
- Palm dipped (Fortix™ Foam Nitrile)
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121A), EN 16350
- EN 388:2016 (4121A), EN 16350
- EN 388:2016 (4121A), EN 16350
- EN 388:2016 (4121A), EN 16350
- EN 388:2016 (4121A), EN 16350
- EN 388:2016 (4121A), EN 16350
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +
- +
- +
- +