Cut resistant industrial gloves, EDGE® 48-702 series
Supplier: Ansell
Certificates
About this item
Besides providing excellent cut and abrasion protection and a firm grip, especially in dry and slightly oily environments, EDGE® 48-702 microfoam nitrile gloves also ensure higher tactile sensitivity and dexterity compared to foam nitrile gloves.
- Seamless liner made from HPPE, Nylon, Polyester and Spandex
- Coated palm is thinner than most comparable NBR gloves
- Breathable coating keeps hands cool and dry
- Composed of FDA-compliant materials, these safety gloves are suitable for handling and packaging food
Recommended for: Automotive, machinery and equipment, metal fabrication, oil and gas.
Certifications: EN 388:2016 (4X42B); Cat. II
Specifications
- Catalog No:
- ANSE48-702/9
- ANSE48-702/10
- ANSE48-702/11
- ANSE48-702/8
- ANSE48-702/7
- ANSE48-702/6
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- HPPE/nylon/polyester/spandex
- HPPE/nylon/polyester/spandex
- HPPE/nylon/polyester/spandex
- HPPE/nylon/polyester/spandex
- HPPE/nylon/polyester/spandex
- HPPE/nylon/polyester/spandex
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile