Industrial gloves, HyFlex® 11-725
Supplier: Ansell
Certificates
About this item
Comfortable, dexterous medium-duty work gloves, offering users moderate cut resistance.
- INTERCEPT™ Cut Resistance Technology: Moderate cut resistance
- HPPE/Lycra® liner: Heightened levels of comfort and flexibility, enabling workers to maintain productivity throughout the day
- PU coating increases dexterity, while providing users a more secure grip in both dry and slightly oily environments
Recommended for: Assembling metal parts and components; assembly of white goods; metalwork, glass processing; handling sharp or irregular metal pieces; press injection molding.
Certifications: EN 388:2016 (4342B)
Specifications
- Catalog No:
- ANSE11-725/8
- ANSE11-725/9
- ANSE11-725/6
- ANSE11-725/7
- ANSE11-725/10
- ANSE11-725/11
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Liner material:
- HPPE/nylon/spandex
- HPPE/nylon/spandex
- HPPE/nylon/spandex
- HPPE/nylon/spandex
- HPPE/nylon/spandex
- HPPE/nylon/spandex
- Finish:
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Gauge:
- 15
- 15
- 15
- 15
- 15
- 15
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile