HyFlex® 11-757 Cut-Resistant Industrial Gloves with INTERCEPT™ Technology
Supplier: Ansell
Certificates
About this item
Ansell's lightest and thinnest ANSI A7/ISO level F cut resistant touchscreen glove.
- Ultra-lightweight and ultra-thin breathable gloves are touchscreen compatible
- High dexterity combined with unmatched comfort and precision while achieving excellent abrasion resistance
- Tailored fit at the base of the pinky finger better matches natural hand contours
- Dermatologically approved
Extreme cut protection featuring INTERCEPT™ Technology while maintaining enhanced flexibility and tactility. Ultra-thin ANSI cut level A7/ISO cut level F touchscreen compatible glove for enhanced productivity. Ultra-light design provides unmatched dexterity for handling and assembly of very sharp parts. A unique combination of ultra-lightweight comfort and extreme cut protection.
Specifications
- Catalog No:
- ANSE11-757/9
- ANSE11-757/7
- ANSE11-757/6
- ANSE11-757/8
- ANSE11-757/10
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile