Gamma Irradiated Sterility Maintenance Covers, Heat Seal
Supplier: KEYSTONE CLEANROOM PRODUCTS
Certificates
About this item
Ideal for pharmaceutical and biotech production applications.
- Wide range of sizes
- Works well with impulse and rotary heat sealers
- No tools required to open
- Impervious to liquid sanitisation agents
- Visibility inside the bag allows for identification
- Prolong the storage and shelf-life of products sterilised 'in-house'
After unloading the autoclave, under Grade A LAF, place wrapped, steam sterilised items into sterility maintenance bags and seal closed. This technique captures the conditions of Grade A and protects sterilised products from external sources of contamination during storage and transport through lesser classified areas.
Certifications: Certificates of Irradiation and Certificates of Conformance sent with shipments.
Packaging: The sterility maintenance covers are packaged in Tyvek Chevron peel pouches with Lot # and expiration date for traceability.