Benchtop Freeze Drying System Alpha 1-4 LSCplus universal package with RV8 Vacuum Pump
Catalog # CHRS102141-1
Supplier: CHRIST MARTIN
Specifications
- Description:Alpha 1-4 LSCplus universal package with vacuum pump RV8
- Pk:2
Specifications
About this item
The Alpha 1-4 LSCplus universal package with vacuum pump RV8. The freeze-drying package includes everything you need to use.
- Benchtop Alpha 1-4 LSCplus freeze-drying system, 230 V, 50 Hz, air-cooled, 0.51 kW refrigeration machine, CFC- and HCFC-free refrigerant with integrated base plate for directly attaching drying chambers or accessories with a diameter of 300 mm , Table top, ice condenser, ice condenser chamber and defrost water and ventilation valve made entirely of stainless steel, ice capacity max. 4 kg, ice condenser output max. 3 kg/24 h, ice condenser temperature approx. -55 °C, ice condenser chamber 6.5 litres, equipped with hot gas defrost, vertical arrangement, suitable for the use of accessories for freezing and drying in a range from approx. -25 to +60 °C, including vacuum sensor
- Equipped with LSCplus system control (Lyo-Screen-Control plus): VGA-TFT color display, graphics-oriented user interface with touchscreen, clear, intuitive user interface with quick access to essential process data, graphic display of drying programs as known from the Christ LPCplus software, data storage on USB (optional), multilingual user interface, password protection, display of maintenance intervals
- Extension unit HED-16 LSCplus for expanding the systems Alpha 1-4 LSCplus and Alpha 2-4 LSCplus and Beta 1-8 LSCplus and Beta 2-8 LSCplus for the electrical heating of shelves, as well as for measuring the sample temperature including product sensor
- Vacuum tubing including standard flange connections DN 25/25 ISO-KF for external connection of the vacuum pump
- WST base plate for Alpha LSCplus, Beta LSCplus, Gamma LSCplus and Delta LSCplus series freeze-drying systems, use when drying outside the ice condenser chamber (two-chamber process)
- 3 unheated stainless steel shelves Ø 265 mm, distance between shelves 79 mm, drying chamber are required
- Acrylic glass drying chamber Ø 300 mm, clear height 295 mm, for holding plate racks, 12 connections for rubber valves, external dimensions: Ø×height: 300×
- 335 mm
- Rubber valve for connecting vessels, with DIN internal ground joint NS 29/32 or distributor
- RV8 vacuum pump 115-230V, 50/60 Hz, includes oil mist filter has a pumping speed of 8.5 m³/h, including small flange connections, vacuum tubing is required