Order Entry
Germany
ContactUsLinkComponent
J.T.Baker® Endonuclease
J.T.Baker® Endonuclease
Supplier:  Avantor
Certificates
undefined
J.T.Baker® Endonuclease
Supplier:  Avantor
Certificates

Specifications

  • CAS No.:
    9025-65-4
  • Storage conditions:
    -20 °C
  • Enzyme name:
    Endonuclease
  • Grade:
    Ultrapure Bioreagent

Specifications