Order Entry
Germany
ContactUsLinkComponent
1,3-Dimethyl-2-imidazolidinone ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Corrosive
Health
Irritant
1,3-Dimethyl-2-imidazolidinone ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Catalog # 8.18212.6190
Supplier:  Merck
CAS Number:  
Corrosive
Health
Irritant
1,3-Dimethyl-2-imidazolidinone ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Catalog # 8.18212.6190
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Steel barrel
  • Pk:
    190 L
  • Seq:
    5514

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    8.2 °C
  • Molar Mass:
    114.14 g/mol
  • Boiling Point:
    104 °C (20 hPa)
  • Vapor Pressure:
    < - 1 hPa (20 °C)
  • Flash Point:
    114 °C
  • Refractive Index:
    1.472 (20 °C, 589 nm)
  • Explosion Limit:
    1.3 - 8.4 %(V)
  • Density:
    1.056 g/cm3 (20 °C)
  • pH:
    (H2O) neutral
  • Ignition Point:
    305 °C
  • Synonyms:
    Dimethylethylene urea, DMI, DMEU
  • Hill Formula:
    C5H10N2O

Specification Test Results

About this item

 :
 :114.15 g/mol
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS08
 :GHS07
CAS Number:80-73-9
Formula:C₅H₁₀N₂O
MDL Number:MFCD00003188