Order Entry
Germany
Orders LinkContactUsLinkComponent
Graphite

Graphite

Bottle, 1 kg
Catalog # SIAL282863-1KG
Supplier:   Merck
Graphite
Catalog # SIAL282863-1KG
Supplier:  Merck
CAS Number:  7782-42-5
Graphite

Bottle, 1 kg

CAS Number:  7782-42-5
Price
72,70 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7782-42-5
  • Formula:C
  • MDL Number:MFCD00144065
  • Molecular Weight:12.01 g/mol
  • Merck Index:12,04560
  • Melting Pt:3827 °C
  • Density:2.09…2.23 g/cm³ (20 °C)
  • EINECS:231-955-3
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.