Order Entry
Germany
ContactUsLinkComponent
Ammonium hydrogen carbonate ≥99.0%, ReagentPlus®
Irritant
Ammonium hydrogen carbonate ≥99.0%, ReagentPlus®
Catalog # SIALA6141-25G
Supplier:  Merck
CAS Number:  
Irritant
Ammonium hydrogen carbonate ≥99.0%, ReagentPlus®
Catalog # SIALA6141-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :79.06 g/mol
Merck Index:13,00497
Melting Pt:35 °C
Density:1.57 g/cm³ (20 °C)
EINECS:213-911-5
 :GHS07
CAS Number:1066-33-7
Formula:NH₄HCO₃
MDL Number:MFCD00012138