Order Entry
Germany
ContactUsLinkComponent
Calcium chloride dihydrate ≥99.0% FCC
Calcium chloride dihydrate ≥99.0% FCC
Catalog # SPCMC1099-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Calcium chloride dihydrate ≥99.0% FCC
Catalog # SPCMC1099-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    99.0 - 107.0 %
  • Arsenic (As):
    max. 3 mg/kg
  • Fluoride:
    max. 0.004 %
  • Lead (Pb):
    max. 5 mg/kg
  • Magnesium and Alkali Salts:
    max. 4.0 %

Specification Test Results

About this item

 :
 :147.01 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:10035-04-8
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613