Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Creatinine NF

Supplier:   Spectrum Chemical
Creatinine NF
Supplier:  Spectrum Chemical
Certificates

4 Options Available Below

Product Details & Documents

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-466-7
Specifications for Product Family

Specification Test Results

  • Assay:
    98.5% Min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 107,00 €
    107,00 €
    1 * 25 g
       107,00 €

  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 290,00 €
    290,00 €
    1 * 100 g
       290,00 €

  • Poly pail
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 200,00 €
    1 200,00 €
    1 * 1 kg
       1 200,00 €

  • Poly pail
    12 kg
    Log in for availability
    Log in for availability
    1 * 12 kg
    1 * 12 kg - 4 480,00 €
    4 480,00 €
    1 * 12 kg
       4 480,00 €