Potassium carbonate, anhydrous ≥99.5%, granular USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:584-08-7
:
Formula:K₂CO₃
MDL Number:MFCD00011382
:138.21 g/mol
Merck Index:13,07702
Melting Pt:891 °C
Density:2.42 g/cm³ (20 °C)
EINECS:209-529-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:99.5% Min.