Utility media, HyClone SFM4CHO
Catalog #
Product Details & Documents
HyClone™ SFM4CHO-Utility media is a serum- and protein-free high performance media designed for growth of many CHO cells with minimal adaptation.
- Designed for high cell yield and recombinant protein production
- Supports multiple CHO clones
- Allows for direct or sequential adaptation
- Supports growth in multiple cell culture systems
- Manufactured from traceable components according to cGMP guidelines
HyClone SFM4CHO-Utility is a versatile cell culture medium designed to support the growth of multiple Chinese hamster ovary (CHO) cell clones and production of a variety of recombinant proteins. The medium enables excellent growth of many CHO cells with minimal adaptation.
Dokumente
GE Hyclone CHO flyer SFM4CHO Utility