MIU medium base dehydrated culture media
Catalog # HIMEM1076-500G
Supplier: HIMEDIA
CAS Number:
Specifications
- Pk:500 g
- Description:MIU medium base
- Seq:01
Specifications
About this item
For the detection of motility and urease and indole production.
- Microbiological culture media
- 500 g poly bottles