Dey-Engley neutralising broth base dehydrated culture media
Catalog # HIMEM187-500G
Supplier: HIMEDIA
CAS Number:
Specifications
- Pk:500 g
- Description:Dey-Engley neutralizing broth base
Specifications
About this item
Used along with Dey-Engley neutralising broth base for neutralizing and determining bactericidal activity of quaternary ammonium compounds.
- Microbiological culture media
- 500 g poly bottles