Order Entry
Germany
ContactUsLinkComponent
Antibiotic assay medium no. 8 dehydrated culture media
Antibiotic assay medium no. 8 dehydrated culture media
Supplier:  HIMEDIA
Certificates
Antibiotic assay medium no. 8 dehydrated culture media
Supplier:  HIMEDIA

Specifications

  • Storage temperature:
    15…25 °C
  • Application:
    Microbiological assay of mitomycin, plicamycin, and vancomycin in accordance with USP.

Specifications