Pierce™ Digestion Indicator for Mass Spectrometry, Thermo Scientific
Catalog # PIER84841
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 µG
- Description:Pierce™ Digestion Indicator for Mass Spectrometry
- Seq:01
- Storage conditions:store at -20˚C. Product is shipped on dry ice.
Specifications
About this item
The Digestion Indicator for Mass Spectrometry is a non-mammalian recombinant protein (26kDa) with five signature peptides for use in determining the digestion efficiency and reproducibility across multiple samples.
- Versatile
- For use with mass spectrometer
- Non-mammalian protein source
This product is sufficient for production of five signature peptides upon digestion for mass spectrometry.
Caution: For Research Use Only. Not for use in diagnostic procedures.