Capping Reagent 84/16 Tetrahydrofuran/N-Methylimidazole (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR650-4X2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 L
- Seq:5114
- Pk Info:4x 2.5 L
Specifications
About this item
:
Density:0.910 g/cm3 (25 °C)
Flash Pt:–15 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS08
:GHS02