Capping Reagent, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Supplier: Honeywell Chemicals
Certificates
About this item
:
Density:0.881 g/cm3 (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:2924
ADR:3,II
:GHS02
:GHS08
:GHS05
:GHS07
Specification Test Results
- Appearance:complying
- Identity (IR):complying
- Assay Of Acetonitrile:49.0 - 51.0 Vol.%
- Assay Of 2,6-Lutidine:29.0 - 31.0 Vol.%
- Acetic aNHydride (GC):19.0 - 21.0 Vol.%
- Water (Karl Fischer):Max. 50 ppm
- Colour (Apha) At Time Of Packaging:Max. 30