Acetic acid 100%, meets analytical specifications of Reagent European Pharmacopoeia, BP, USP, FCC, E260, Burdick & Jackson™
Catalog # HONC65923-200L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:200 L
- Seq:5549
Specifications
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1.013 hPa)
Melting Pt:16 °C
Density:1.05 g/cm3 (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152