Order Entry
Germany
ContactUsLinkComponent
 
Thymolphthalein solution for PANPEHA indicator paper, Riedel-de Haën™
Catalog # HONC34663-5L
CAS Number:  
Thymolphthalein solution for PANPEHA indicator paper, Riedel-de Haën™
Catalog # HONC34663-5L
CAS Number:  

Specifications

  • Pack type:
    Plastic can
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :430.54 g/mol
CAS Number:125-20-2
Formula:C₂₈H₃₀O₄
MDL Number:MFCD00005909