Order Entry
Germany
ContactUsLinkComponent
Acetone, Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Irritant
Flammable
Acetone, Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Catalog # HONC24201B-500L
   Safer Human and Environmental Health
CAS Number:  
Irritant
Flammable
Acetone, Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Catalog # HONC24201B-500L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Stainless steel IBC, returnable
  • Pk:
    500 L
  • Seq:
    5675
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

About this item

 :
 :58.08 g/mol
Boiling Pt:55…57 °C (1,013 hPa)
Melting Pt:–95 °C
Density:0.79 g/cm3 (20 °C)
Flash Pt:–17 °C
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
 :GHS07
 :GHS02
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765