Surfactant, Tergitol™ 15-S-9
Supplier: Spectrum Chemical
Certificates
About this item
Tergitol™ 15-S-9, Surfactant is a non-ionic secondary alcohol ethoxylate surfactant and is an excellent emulsifier and detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints and coatings.
- Superior detergency
- Soluble in water
- Rapid dissolution and good rinseability
- Low odour
- Narrow gel range
- Excellent formulation and handling properties
Density at 20 °C (68 °F): 1,006 g/ml.