Capping Reagent 80/20 Acetonitrile/NMI (N-Methylimidazole) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR654-4X2.5GL
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle, GL45
- Pk:10 L
- Seq:5114
- Pk Info:4x 2.5 L
Specifications
About this item
:
Boiling Pt:86.6 °C (main component)
Melting Pt:–43.8 °C (main component)
Density:0.827 g/cm³ (20 °C)
Flash Pt:6 °C (main component)
Storage Temperature:Ambient
EINECS:200-835-2
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS08
:GHS02