Tetrahydrofuran, Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Catalog # HONC72187-45L-RC
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Stainless steel drum, returnable
- Pk:45 L
- Seq:5372
Specifications
Specification Test Results
- Assay (GC):Min. 99.8 %
- Density (20°C):0.886 - 0.888 g/ml
- Refractive index (n 20/D):1.406 - 1.408
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Potassium (K):Max. 0.00005 %
- Lithium (Li):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Molybdenum (Mo):Max. 0.00001 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Strontium (Sr):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- Peroxides (as H₂O₂):Max. 0.005 %
- Assay BHT (GC):200 - 300 ppm
- Appearance (color):complying
- Appearance (form):complying
- Mixable with H₂O:complying
- Mixable with ethanol:complying
- APHA:Max. 20
Specification Test Results
About this item
:
:72.11 g/mol
Boiling Pt:65 °C (1,013 hPa)
Melting Pt:–108 °C
Density:0.89 g/cm3 (20 °C)
Flash Pt:–21 °C
Storage Temperature:Ambient
UN:2056
ADR:3,II
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356